PUREGRIP® Round Glass Media Storage Bottles with GL 45 VersaCap®
Supplier: Foxx Life Sciences
1501016-FLS 1501021-FLS 1501024-FLS 1501029-FLS 1501030-FLS 1501031-FLS 1501033-FLS 1501038-FLS 1501040-FLS 1519016-FLS 1519021-FLS 1519024-FLS 1519029-FLS 1519030-FLS 1519031-FLS 1519033-FLS 1519038-FLS 1519040-FLS
76379-736CS 154.84 USD
76379-736 76379-738 76379-740 76379-742 76379-744 76379-746 76379-748 76379-750 76379-752 76379-754 76379-756 76379-758 76379-760 76379-762 76379-764 76379-766 76379-768 76379-770
PUREGRIP® Round Glass Media Storage Bottles with GL 45 VersaCap®
Bottles Glass Bottles
PUREGRIP® reusable round glass media storage bottles with GL 45 caps are available in clear or amber finishes.
  • 3.3 borosilicate glass material
  • 100 ml to 20 L volumes available
  • Polypropylene cap material offers the highest cap temperature rating in the industry, at 142 °C
  • Clear or amber finishes available
  • With white enamel graduations
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
PUREGRIP® Round Glass Media Storage Bottles are made from 3.3 borosilicate glass with the patented advantages of the new GL 45 VersaCap®, a leap forward in laboratory glassware. Ideal for mixing, storage, and transportation of media, buffers, chemicals, and solvents, PUREGRIP® bottles offer versatility across a wide variety of laboratory needs. With Foxx's revolutionary GL 45-threaded polypropylene VersaCap®, these bottles are substantially easier to hold securely, improving lab safety.
Certificaciones: Class/quality grade: Type 1, Class A .