NEX S Fume Extractor Arms, Nederman
Supplier: Nederman
10520232 10521232 10520332 10521332 10520432 10521432
46102-202EA 9962.25 USD
46102-202 46102-204 46102-206 46102-208 46102-210 46102-212
NEX S Fume Extractor Arms, Nederman
Fume Extractors
Ideal for environments containing explosive dust or gas and applications with high hygienic demands
Units feature high suction capacity, mobility, and ease of maintenance. Hoses are mounted parallel to self-supporting external flexible arms by straps and are easily detached for cleaning. Interior of hose is free from components allowing for minimal dust and particle accumulation, maximum airflow, minimal pressure drop, and low noise level. Arms are made of acid-proofed polished stainless steel and provide light and precise maneuverability, while remaining stable when positioned. Units are equipped with a horizontally pivoted boom allowing for extra reach. The multipositioned hood, rectangular flange, and protection grille improve capture capacity. All profiles are completely sealed and all threads are covered by cap nuts. Units have a double-grounded system with an antistatic grounded ventilation hose that prevents static electricity and sparks.
Información para pedidos: Fume extractor arms are not supplied with fans. For compatible fans and starters, see 46102-214 series.