Adson Forceps, Straight
Supplier: WORLD PRECISION INSTRUMENTS LLC
503719-12 503720-12
77663-386PK 56.98 USD
77663-386 77663-388
Adson Forceps, Straight
Forceps
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Envase: Package of 12.