Gold ≥99.99%, disk, as rolled, Diameter 32.75 mm

Supplier: Goodfellow

1000046570 1000047533
77681-688EA 2554.65 USD
77681-688 77681-911
Gold ≥99.99%, disk, as rolled, Diameter 32.75 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR