Gold ≥99.99%, disk, as roll, Diameter 18.7 mm
Supplier: GOODFELLOW CORPORATION
1000046511
1000047462
77681-660EA
1111.23
USD
77681-660
77681-883
Gold ≥99.99%, disk, as roll, Diameter 18.7 mm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...