Gold ≥99.99%, disk, as roll, Diameter 7.8 mm
Supplier: GOODFELLOW CORPORATION
1000046465
1000047408
77681-639EA
360.6
USD
77681-639
77681-862
Gold ≥99.99%, disk, as roll, Diameter 7.8 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...