Gold ≥99.99%, disk, as roll, Diameter 5.9 mm
Supplier: GOODFELLOW CORPORATION
1000046449
1000047392
77681-632EA
272.1
USD
77681-632
77681-855
Gold ≥99.99%, disk, as roll, Diameter 5.9 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...