Gold ≥99.99%, disk, as rolled, Diameter 25.4 mm
Supplier: Goodfellow
1000030559
1000031912
1000030693
1000030661
1000031565
1000032075
77680-819EA
392.81
USD
77680-819
77681-130
77680-861
77680-848
77681-042
77681-172
Gold ≥99.99%, disk, as rolled, Diameter 25.4 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...