Gold ≥99.99%, disk, as roll, Diameter 20 mm
Supplier: GOODFELLOW CORPORATION
1000030557
1000030692
1000032073
1000030659
1000031562
1000031911
77680-818EA
304.49
USD
77680-818
77680-860
77681-171
77680-847
77681-041
77681-129
Gold ≥99.99%, disk, as roll, Diameter 20 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...