Gold ≥99.95%, disk, as rolled, Diameter 45 mm

Supplier: Goodfellow

1000019329 1000044127 1000043630 1000047070
77680-717EA 4274.34 USD
77680-717 77681-410 77681-337 77681-849
Gold ≥99.95%, disk, as rolled, Diameter 45 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR