Gold ≥99.95%, disk, as roll, Diameter 36.7 mm
Supplier: GOODFELLOW CORPORATION
1000046830
1000044113
1000043614
1000047052
77681-767EA
3116.56
USD
77681-767
77681-403
77681-330
77681-842
Gold ≥99.95%, disk, as roll, Diameter 36.7 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...