Gold ≥99.95%, disk, as rolled, Diameter 23.5 mm
Supplier: Goodfellow
1000043573
1000044067
1000046782
1000046985
77681-309EA
676.39
USD
77681-309
77681-385
77681-747
77681-821
Gold ≥99.95%, disk, as rolled, Diameter 23.5 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...