Gold ≥99.95%, disk, as rolled, Diameter 18.7 mm
Supplier: Goodfellow
1000044043
1000046740
1000043553
1000046951
77681-373EA
788.53
USD
77681-373
77681-735
77681-298
77681-809
Gold ≥99.95%, disk, as rolled, Diameter 18.7 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...