Gold ≥99.95%, disk, as roll, Diameter 7.5 mm

Supplier: GOODFELLOW CORPORATION

1000043989 1000043504 1000046666 1000046881
77681-349EA 271.08 USD
77681-349 77681-274 77681-711 77681-785
Gold ≥99.95%, disk, as roll, Diameter 7.5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR