Gold ≥99.95%, disk, as roll, Diameter 33 mm
Supplier: GOODFELLOW CORPORATION
1000046824
1000021900
1000043606
1000047040
77681-764EA
2647.22
USD
77681-764
77680-720
77681-326
77681-838
Gold ≥99.95%, disk, as roll, Diameter 33 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...