Gold ≥99.999%, disk, as roll, Diameter 24 mm

Supplier: Goodfellow

1000044940 1000041462 1000046346 1000044247
77681-531EA 3871.12 USD
77681-531 77681-252 77681-600 77681-458
Gold ≥99.999%, disk, as roll, Diameter 24 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR