Gold ≥99.999%, disk, as roll, Diameter 22.75 mm

Supplier: GOODFELLOW CORPORATION

1000028339 1000041455 1000044937 1000046339
77680-730EA 1610.6 USD
77680-730 77681-249 77681-528 77681-597
Gold ≥99.999%, disk, as roll, Diameter 22.75 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR