Gold ≥99.999%, disk, as rolled, Diameter 20.7 mm
Supplier: Goodfellow
1000041449
1000044236
1000044931
1000046332
77681-246EA
5256.46
USD
77681-246
77681-453
77681-525
77681-594
Gold ≥99.999%, disk, as rolled, Diameter 20.7 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...