Gold ≥99.999%, disk, as roll, Diameter 14.4 mm
Supplier: GOODFELLOW CORPORATION
1000044206
1000041414
1000044902
1000046291
77681-439EA
834.46
USD
77681-439
77681-232
77681-511
77681-582
Gold ≥99.999%, disk, as roll, Diameter 14.4 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...