Gold ≥99.999%, disk, as roll, Diameter 11.125 mm
Supplier: GOODFELLOW CORPORATION
1000046272
1000041393
1000044186
1000044886
77681-574EA
431.98
USD
77681-574
77681-223
77681-430
77681-503
Gold ≥99.999%, disk, as roll, Diameter 11.125 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...