Gold ≥99.999%, disk, as roll, Diameter 11 mm

Supplier: GOODFELLOW CORPORATION

1000046271 1000041390 1000044884 1000044184
77681-573EA 426.3 USD
77681-573 77681-222 77681-502 77681-429
Gold ≥99.999%, disk, as roll, Diameter 11 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR