Gold ≥99.999%, disk, as roll, Diameter 7.7 mm
Supplier: GOODFELLOW CORPORATION
1000041376
1000044873
1000044170
1000046255
77681-215EA
1215.01
USD
77681-215
77681-496
77681-422
77681-566
Gold ≥99.999%, disk, as roll, Diameter 7.7 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...