Gold ≥99.999%, disk, as rolled, Diameter 7.5 mm
Supplier: Goodfellow
1000041374
1000044870
1000044169
1000046252
77681-214EA
1171.91
USD
77681-214
77681-495
77681-421
77681-565
Gold ≥99.999%, disk, as rolled, Diameter 7.5 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...