Gold ≥99.999%, disk, as roll, Diameter 4.75 mm
Supplier: GOODFELLOW CORPORATION
1000044151
1000041357
1000046234
1000044848
77681-413EA
236.93
USD
77681-413
77681-205
77681-558
77681-487
Gold ≥99.999%, disk, as roll, Diameter 4.75 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...