Gold ≥99.999%, disk, as rolled, Diameter 6 mm

Supplier: Goodfellow

1000031149 1000030348 1000031245 1000031884 1000031401
77680-968EA 292.23 USD
77680-968 77680-756 77680-991 77681-124 77681-024
Gold ≥99.999%, disk, as rolled, Diameter 6 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR