Gold ≥99.999%, disk, as roll, Diameter 20 mm
Supplier: GOODFELLOW CORPORATION
1000030339
1000031122
1000031387
1000031869
1000031224
77680-751EA
4983.64
USD
77680-751
77680-959
77681-018
77681-115
77680-982
Gold ≥99.999%, disk, as roll, Diameter 20 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...