Gold ≥99.9%, disk, Diameter 50.8 mm

Supplier: GOODFELLOW CORPORATION

1000030320 1000030421 1000030375 1000018125 1000014808 1000031748 1000031832 1000030820 1000031640 1000031789
77680-742EA 752.73 USD
77680-742 77680-783 77680-768 77680-714 77680-706 77681-081 77681-106 77680-895 77681-061 77681-092
Gold ≥99.9%, disk, Diameter 50.8 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR