Gold ≥99.95%, disk, as roll, Diameter 40 mm
Supplier: Goodfellow
1000030749
1000030884
1000031076
1000031495
1000031997
1000032107
1000030629
1000031284
1000031950
77680-878EA
1607.04
USD
77680-878
77680-908
77680-948
77681-032
77681-160
77681-189
77680-836
77681-000
77681-146
Gold ≥99.95%, disk, as roll, Diameter 40 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...