Gold ≥99.95%, disk, as roll, Diameter 25 mm
Supplier: Goodfellow
1000030740
1000030878
1000002782
1000031068
1000031994
1000031280
1000032104
1000030624
1000031704
1000031945
77680-875EA
895.78
USD
77680-875
77680-906
77680-692
77680-945
77681-158
77680-998
77681-187
77680-833
77681-068
77681-143
Gold ≥99.95%, disk, as roll, Diameter 25 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...