Fire Polished Glass Capillaries
Supplier: WORLD PRECISION INSTRUMENTS LLC
504949 504950
77663-800EA 127.83 USD
77663-800 77663-802
Fire Polished Glass Capillaries
Pipettes Microcapillaries
Fire polished glass capillaries for use with the Nanoliter 2010 or the older Nanoliter 2000.
  • 300" vial
  • For use with the Nanoliter 2010 or the older Nanoliter 2000
  • Fire polished
  • Compatible with Nanoliter 2020 and Nanoliter 2020 injector head
Fire-Polished glass capillaries are easier to insert into microelectrode holders without damaging the gasket. More importantly, firepolished glass won’t scratch the chloridized wire used in a recording electrode. Fire-polishing does not affect the glass’s mechanical or electrical properties.