Human Recombinant SCF (CTGrade GMP) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
500-02-100UG 500-02-1MG 500-02-50UG
77660-950EA 1421.4 USD
77660-950 77660-952 77660-954
Human Recombinant SCF (CTGrade GMP) (from E. coli)
Proteins and Peptides
SCF is a multipotent factor which affects cells of the myeloid, erythroid, mast, and lymphoid lineages.
  • High quality
  • Low endotoxin
  • Lot-to-lot consistency
  • Manufactured following GMP guidelines
Exists both as a membrane-bound and soluble form
Expressed by fibroblasts and endothelial cells promoting proliferation, migration, survival, and differentiation in hematopoietic progenitors, melanocytes, and germ cells
Promotes mast cell secretory function, chemotaxis, and adhesion
Enhances secretion of mediators in response to IgE-dependent activation
The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance
Host Cell Protein: ≤30 ng/ml
Host Cell DNA: ≤0.01 ng/ml
Sterility testing: No Growth
Mycoplasma: Negative
Precaución: For research use or further manufacturing only.