PROPOR BR MURUS Capsules
Supplier: PARKER HANNIFIN TE
ZLBRK-020BB-PNL ZLBRK-020BB-PSL ZLBRK-020TT-PNL ZLBRK-020TT-PSL ZLBR1-020TT-PNL ZLBR1-020TT-PSL ZLBR2-020BB-PNL ZLBR2-020BB-PSL ZLBR2-020TT-PNL ZLBR2-020TT-PSL ZLBR3-020BB-PNL ZLBR3-020BB-PSL ZLBR3-020TT-PNL ZLBR3-020TT-PSL
76924-080EA 393.82 USD
76924-080 76924-094 76924-074 76924-088 76924-068 76924-082 76924-076 76924-090 76924-070 76924-084 76924-078 76924-092 76924-072 76924-086
PROPOR BR MURUS Capsules
Filters Capsule Filters
PROPOR BR filters have been specifically designed for the fast and cost-effective bioburden reduction of pharmaceutical solutions.
  • Brevundimonas diminuta retention of LRV >5 for efficient bioburden reduction
  • Additional prefilter layer gives excellent throughput to blockage
  • Low binding for minimal product loss
PROPOR BR filters feature an integral meltblown prefilter layer to maximize dirt holding capacity while the polyethersulphone membrane guarantees a bioburden log reduction of greater than 5, giving excellent microbial protection. This makes PROPOR BR filters ideal for bioburden reduction of LVPs prior to terminal sterilization.
PROPOR BR filters are also ideally suited to prefiltration and bioburden reduction prior to sterilizing grade membrane filters. The robust construction of PROPOR BR filters guarantees consistent performance on multiple batches.
PFAS-free options available.
Envase: Supplied with protection cups on vent and drain ports.