Backhaus Towel Clamps
Supplier: WORLD PRECISION INSTRUMENTS LLC
501320 501700
77662-582EA 56.98 USD
77662-582 77662-658
Backhaus Towel Clamps
Forceps
Backhaus towel clamps, also known as the Backhaus forceps or towel forceps, are designed for securing and attaching surgical drapes or towels to a patient's skin or other surfaces. The Backhaus towel clamp has a scissor-like design with ring handles. These curved, pointed jaws have small, sharp, and inward-facing tips for a secure grip when the instrument is closed. A ratchet mechanism lets you securely lock them in a closed position to maintain a firm grip on the drapes.
  • 9 cm (3.5") or 13 cm (5.25") Long
  • Curved, pointed tips
  • Locking handle
  • Surgical grade stainless steel
Choose the size based on the thickness of the material to be clamped and the area of application. These surgical forceps are both of which are autoclavable. This is recommend for properly cleaning all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.