(2R,5S)-tert-Butyl 2,5-diethylpiperazine-1-carboxylate 97%, Ambeed.Inc

Supplier: Ambeed

Synonyms: tert-butyl (2R,5S)-2,5-diethylpiperazine-1-carboxylate, (2R,5S)-1-Boc-2,5-diethyl-piperazine

A915345-1G A915345-250MG A915345-5G
77684-870EA 447 USD
77684-870 77684-871 77893-717
(2R,5S)-tert-Butyl 2,5-diethylpiperazine-1-carboxylate 97%, Ambeed.Inc
(2R,5S)-tert-Butyl 2,5-diethylpiperazine-1-carboxylate

Precaución: For research use only.

Formula: C₁₃H₂₆N₂O₂
MDL Number: MFCD08686670
CAS Number: 1240588-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR