3-Chloro-2,4-difluorobenzylamine 95%, Ambeed.Inc

Supplier: Ambeed
Danger

A591416-100MG A591416-1G A591416-250MG A591416-5G
77684-458EA 50 USD
77684-458 77886-432 77886-433 77886-434
3-Chloro-2,4-difluorobenzylamine 95%, Ambeed.Inc
3-Chloro-2,4-difluorobenzylamine

Precaución: For research use only.

Formula: C₇H₆ClF₂N
MDL Number: MFCD06660335
CAS Number: 886761-65-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR