Di-tert-butyl(2',4',6'-triisopropyl-4-methoxy-3,5,6-trimethyl-[1,1'-biphenyl]-2-yl)phosphine 98%, Ambeed.Inc

Supplier: Ambeed

Synonyms: 2-Di-t-butylphosphino-5-methoxy-3,4,6-trimethyl-2′,4′,6′-tri-i-propylbiphenyl], Me3(ome)tbuxphos

A388699-1G A388699-250MG A388699-5G
77684-120EA 296 USD
77684-120 77886-029 77886-030
Di-tert-butyl(2',4',6'-triisopropyl-4-methoxy-3,5,6-trimethyl-[1,1'-biphenyl]-2-yl)phosphine 98%, Ambeed.Inc
Di-tert-butyl(2',4',6'-triisopropyl-4-methoxy-3,5,6-trimethyl-[1,1'-biphenyl]-2-yl)phosphine

Precaución: For research use only.

Formula: C₃₃H₅₃OP
MDL Number: MFCD22666028
CAS Number: 1359986-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR