Mouse Recombinant FLT-3 Ligand (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-06AF-100UG 200-06AF-10UG 200-06AF-1MG 200-06AF-500UG
77660-814EA 601.52 USD
77660-814 77660-816 77660-818 77660-820
Mouse Recombinant FLT-3 Ligand (Animal free) (from E. coli)
Proteins and Peptides
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation. FLT-3 ligand signaling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor. FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3).
  • High Quality
  • Low Endotoxin
  • Affordable
Stability: 12 months from date of receipt when stored at −20 to −80 °C as supplied. 1 month when stored at 4 °C after reconstituting as directed. 3 months when stored at −20 to −80 °C after reconstituting as directed.
Precaución: For research use only.