ATC 0175 hydrochloride ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N-[cis-4-[[4-(Dimethylamino)-2-quinazolinyl]amino]cyclohexyl]-3,4-difluorobenzamide hydrochloride

A287527-10MG A287527-25MG
ALA287527-10MGEA 199.41 USD
ALA287527-10MG ALA287527-25MG
ATC 0175 hydrochloride ≥98% (by HPLC)
ATC 0175 hydrochloride
Formula: C₂₃H₂₅F₂N₅O·HCl
MW: 461.94 g/mol
Storage Temperature: Refrigerator
CAS Number: 510733-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8e556ab1658a96a354ee3293e6aa47cb39345ae&vid=f8e556ab1658a96a354ee3293e6aa47cb39345ae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR