ABT 724 trihydrochloride ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A287403-10MG
A287403-25MG
A287403-5MG
ALA287403-10MGEA
157.9
USD
ALA287403-10MG
ALA287403-25MG
ALA287403-5MG
ABT 724 trihydrochloride ≥98%
ABT 724 trihydrochloride
Product Describtion:ABT-724 is a dopamine D4 receptor agonist. ABT-724 has been used in the study of impulsive behavior and penile erection in rats.
Formula:
C₁₇H₂₂Cl₃N₅ Storage Temperature: Refrigerator |
CAS Number:
587870-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...