5-((5-(2,8-Dimethyl-5H-dibenzo[a,d][7]annulen-5-yl)-2-oxo-4-thioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)-N-(1H-tetrazol-5-yl)furan-2-carboxamide ≥97% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 5-[[5-(2,8-Dimethyl-5H-dibenzo[a,d]cyclohepten-5-yl)-3,4-dihydro-2-oxo-4-thioxo-1(2H)-pyrimidinyl]methyl]-N-2H-tetrazol-5-yl-2-furancarboxamide, AR-C118925XX, AR-C 118925XX

A287202-1MG A287202-5MG
ALA287202-1MGEA 173.76 USD
ALA287202-1MG ALA287202-5MG
5-((5-(2,8-Dimethyl-5H-dibenzo[a,d][7]annulen-5-yl)-2-oxo-4-thioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)-N-(1H-tetrazol-5-yl)furan-2-carboxamide ≥97% (by HPLC), Moligand™
5-((5-(2,8-Dimethyl-5H-dibenzo[a,d][7]annulen-5-yl)-2-oxo-4-thioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)-N-(1H-tetrazol-5-yl)furan-2-carboxamide
Formula: C₂₈H₂₃N₇O₃S
MW: 537.59 g/mol
Storage Temperature: Ambient
CAS Number: 216657-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac17d46fbd5d4a6faf427b6f8180de638559d3ae&vid=ac17d46fbd5d4a6faf427b6f8180de638559d3ae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR