AM 4113 ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide, CB1 antagonist 2
A287160-100MG
A287160-10MG
A287160-250MG
A287160-50MG
A287160-5MG
ALA287160100MGEA
752.31
USD
ALA287160100MG
ALA287160-10MG
ALA287160250MG
ALA287160-50MG
ALA287160-5MG
AM 4113 ≥98% (by HPLC)
AM 4113
Formula:
C₁₇H₁₂Cl₃N₃O MW: 380.66 g/mol Storage Temperature: Freezer |
CAS Number:
614726-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...