(S)-2-Methyl-3-(pyrrolidin-2-ylmethoxy)pyridine dihydrochloride ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: ABT 089 dihydrochloride, Pozanicline 2HCl, 2-Methyl-3-[(2S)-pyrrolidinylmethoxy]pyridine dihydrochloride

A287125-100MG A287125-10MG A287125-25MG A287125-50MG A287125-5MG
ALA287125100MGEA 1175.06 USD
ALA287125100MG ALA287125-10MG ALA287125-25MG ALA287125-50MG ALA287125-5MG
(S)-2-Methyl-3-(pyrrolidin-2-ylmethoxy)pyridine dihydrochloride ≥98% (by HPLC)
(S)-2-Methyl-3-(pyrrolidin-2-ylmethoxy)pyridine dihydrochloride
Formula: C₁₁H₁₈Cl₂N₂O
MW: 265.18 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00929191
CAS Number: 161416-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f375f918c04984c1a0c3dbc491cd81222b5663ac&vid=f375f918c04984c1a0c3dbc491cd81222b5663ac&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR