1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one ≥98% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: AM 0902, 1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-Purin-6-one, AMG0902, 1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one, AM-0902, 1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-purin-6-one

A287071-50MG A287071-5MG A287071-100MG A287071-10MG A287071-25MG
ALA287071-50MGEA 495.81 USD
ALA287071-50MG ALA287071-5MG ALA287071100MG ALA287071-10MG ALA287071-25MG
1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one ≥98% (by HPLC), Moligand™
1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one
Formula: C₁₇H₁₅ClN₆O₂
MW: 370.7930 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31579854
CAS Number: 1883711-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4c0d058d70f7f7f5b75264ba350cb674716bfe5&vid=e4c0d058d70f7f7f5b75264ba350cb674716bfe5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR