AS101 ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Ammonium trichloro[1,2-ethanediolato-O,O']-tellurate, AS 101

A286696-100MG A286696-10MG A286696-25MG A286696-50MG A286696-5MG
ALA286696100MGEA 4313.86 USD
ALA286696100MG ALA286696-10MG ALA286696-25MG ALA286696-50MG ALA286696-5MG
AS101 ≥98%
AS101

Product Introduction:Ossirene (AS101), an immunomodulatory tellurium compound, is a potent IL-1β inhibitor. Ossirene abolishes phosphorylation of STAT3 by inhibiting IL-10. Ossirene potently inhibits Caspase-1 and is used for the autoimmune diseases and certain malignancies.

Formula: C₂H₈Cl₃NO₂Te
MW: 312.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01682758
CAS Number: 106566-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR