AC 5216 ≥98% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Emapunil, NVP-XBD173, XBD173, N-Ethyl-7,8-dihydro-7-methyl-8-oxo-2-phenyl-N-(phenylmethyl)-9H-purine-9-acetamide

A286555-100MG A286555-10MG A286555-25MG A286555-50MG A286555-5MG
ALA286555100MGEA 1246.31 USD
ALA286555100MG ALA286555-10MG ALA286555-25MG ALA286555-50MG ALA286555-5MG
AC 5216 ≥98% (by HPLC), Moligand™
AC 5216
Formula: C₂₃H₂₃N₅O₂
MW: 401.46 g/mol
Storage Temperature: Freezer
CAS Number: 226954-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee87bc829840cc7c0c581765c40a2aa239466c49&vid=ee87bc829840cc7c0c581765c40a2aa239466c49&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR