Ammonium metatungstate hydrate ≥90% WO₃ basis

Supplier: ALADDIN SCIENTIFIC

A283516-100G A283516-25G
ALA283516-100GEA 76.86 USD
ALA283516-100G ALA283516-25G
Ammonium metatungstate hydrate ≥90% WO₃ basis
Ammonium metatungstate hydrate
Formula: (NH₄)₆W₁₂O₃₉·xH₂O
MDL Number: MFCD00042673
CAS Number: 12028-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR