Aluminum hydroxide terephthalate MOF, MIL-53(Al) Surface area 1100-1500 m2/g

Supplier: ALADDIN SCIENTIFIC

A281775-1G A281775-250MG A281775-500MG
ALA281775-1GEA 383.71 USD
ALA281775-1G ALA281775250MG ALA281775500MG
Aluminum hydroxide terephthalate MOF, MIL-53(Al) Surface area 1100-1500 m2/g
Aluminum hydroxide terephthalate

Aluminum terephthalate is an Ultramicroporous metal framework (MOF). Aluminum terephthalate is a microporous metal organic framework (MOF) consisting of 3D Networks with micropores up to 900 PM in diameter. Because it contains octahedral alo4 (OH) 2 units linked by dimethyl 1,4-benzoate ligands, it has the same properties as MIL-53 (AL). It can form a framework with a specific surface area of 1084 2g-1 and a pore volume of about 0.51 cm2g-1.Product ApplicationAluminum terephthalate is mainly used for co2/ch4 and co2/n2 binary gas separation. It can also be used for gas adsorption (hydrogen) and supercritical adsorption of CO2. Separation of C8 alkyl aromatic compounds using aluminum terephthalate.

Formula: C₈H₅AlO₅
Storage Temperature: Ambient
CAS Number: 654061-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR