ABT-263 ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: (R)-4-(4-((4'-Chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((4-((4-morpholino-1-(phenylthio)butan-2-yl)amino)-3-((trifluoromethyl)sulphonyl)phenyl)sulphonyl)benzamide, Navitoclax

A276560-5MG A276560-100MG A276560-25MG A276560-500MG
ALA276560-5MGEA 35.06 USD
ALA276560-5MG ALA276560100MG ALA276560-25MG ALA276560500MG
ABT-263 ≥98%
ABT-263

Shipped at 4°C. Store at -20°C. Store under desiccating conditions.Product descriptionNavitoclax (ABT-263) is a potent and orally active Bcl-2 family protein inhibitor that binds to multiple anti-apoptotic Bcl-2 family proteins, such as Bcl-xL, Bcl-2 and Bcl-w, with a Ki of less than 1 nM.

Formula: C₄₅H₅₅ClF₃N₅O₆S₃
MW: 974.61 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12756219
CAS Number: 923564-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b55536357ea1a043521452f64113818239bc941d&vid=b55536357ea1a043521452f64113818239bc941d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR