AM 580 ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: NSC 608001, CD336, 4-​​[[(5,​​6,​​7,​​8-​​tetrahydro-​​5,​​5,​​8,​​8-​​tetramethyl-​​2-​​naphthalenyl)​carbonyl]​amino]​-​benzoic acid, AM580, 4-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxamido)benzoic acid, RO 40-6055

A276517-10MG A276517-250MG A276517-50MG
ALA276517-10MGEA 93.01 USD
ALA276517-10MG ALA276517250MG ALA276517-50MG
AM 580 ≥98%
AM 580

Product Describtion:AM580 is a retinoic acid receptor agonist with Kd values of 8 nM, 131 nM and 450 nM for RARα, RARβ and RARγ.

Formula: C₂₂H₂₅NO₃
Melting Pt: 260 °C
Storage Temperature: Freezer
MDL Number: MFCD00673916
CAS Number: 102121-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40e722f96e0ef63e4f6b43097e9d230c470b95ba&vid=40e722f96e0ef63e4f6b43097e9d230c470b95ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR