AMG-9810 ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A276243-10MG A276243-25MG A276243-50MG A276243-5MG
ALA276243-10MGEA 140.51 USD
ALA276243-10MG ALA276243-25MG ALA276243-50MG ALA276243-5MG
AMG-9810 ≥99%, Moligand™
AMG-9810

Store at +4°C. Store under desiccating conditions. The product can be stored for up to 12 months.

Formula: C₂₁H₂₃NO₃
Storage Temperature: Freezer
MDL Number: MFCD00955103
CAS Number: 545395-94-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41a90d3718af7dbc9f4c98902ae5a9223d0e9ec1&vid=41a90d3718af7dbc9f4c98902ae5a9223d0e9ec1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR